Amino-PEG9-Amine ≥95%

Supplier: Ambeed
Warning

Synonyms: 3,6,9,12,15,18,21,24,27-Nonaoxanonacosane-1,29-diamine

A753961-1G A753961-250MG A753961-100MG
77622-742EA 150.38 USD
77622-742 77622-744 77783-905
Amino-PEG9-Amine ≥95%
Amino-PEG9-Amine

Precaución: for Research Use Only

Formula: C₂₀H₄₄N₂O₉
MDL Number: MFCD28122963
CAS Number: 474082-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR