6,7-Dihydro-5H-cyclopenta[b]pyridin-5-ol ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A753861-250MG A753861-5G
77684-685EA 39 USD
77684-685 77684-686
6,7-Dihydro-5H-cyclopenta[b]pyridin-5-ol ≥95%, Ambeed.Inc
6,7-Dihydro-5H-cyclopenta[b]pyridin-5-ol

Precaución: For research use only.

Formula: C₈H₉NO
MDL Number: MFCD09800610
CAS Number: 1065609-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR