(1R,4S)-Fmoc-4-aminocyclopent-2-enecarboxylic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (1R,4S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclopent-2-enecarboxylic acid

A753762-1G A753762-250MG
77614-318EA 357.41 USD
77614-318 77614-320
(1R,4S)-Fmoc-4-aminocyclopent-2-enecarboxylic acid ≥98%
(1R,4S)-Fmoc-4-aminocyclopent-2-enecarboxylic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₉NO₄
MW: 349.39 g/mol
MDL Number: MFCD01311756
CAS Number: 220497-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR