5-Bromo-2-thiophenylboronic acid mida ester ≥95%

Supplier: Ambeed
Warning

A753492-5G A753492-25G A753492-250MG A753492-1G
77622-732EA 188.49 USD
77622-732 77783-901 77783-900 77783-899
5-Bromo-2-thiophenylboronic acid mida ester ≥95%
5-Bromo-2-thiophenylboronic acid mida ester

Precaución: for Research Use Only

Formula: C₉H₉BBrNO₄S
MW: 317.9521 g/mol
MDL Number: MFCD11215240
CAS Number: 943552-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR