5-Amino-1-(3,5-dichloro-4-(4-chlorobenzoyl)benzyl)-1H-1,2,3-triazole-4-carboxamide ≥95%

Supplier: Ambeed
Danger

Synonyms: Carboxyamidotriazole

A753409-1MG A753409-5MG A753409-10MG
77618-080EA 10.3 USD
77618-080 77618-082 77686-762
5-Amino-1-(3,5-dichloro-4-(4-chlorobenzoyl)benzyl)-1H-1,2,3-triazole-4-carboxamide ≥95%
5-Amino-1-(3,5-dichloro-4-(4-chlorobenzoyl)benzyl)-1H-1,2,3-triazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₁₇H₁₂Cl₃N₅O₂
MW: 424.67 g/mol
MDL Number: MFCD00866325
CAS Number: 99519-84-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR