N-Methyl-L-serine ≥98%

Supplier: Ambeed
Warning

Synonyms: H-N-Me-Ser-OH, N-Me-Ser-OH

A753348-250MG A753348-1G A753348-100MG A753348-25G
77602-936EA 75.85 USD
77602-936 77674-068 77882-645 77882-646
N-Methyl-L-serine ≥98%
N-Methyl-L-serine

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00037756
CAS Number: 2480-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR