Bis(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98%

Supplier: Ambeed
Danger

Synonyms: Dimesityliodonium triflate, Bis(2,4,6-trimethylphenyl)iodonium triflate

A753078-1G A753078-5G
77573-418EA 45 USD
77573-418 77622-724
Bis(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98%
Bis(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₉H₂₂F₃IO₃S
MW: 514.34 g/mol
MDL Number: MFCD25541454
CAS Number: 139139-80-3
UN: 2923
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR