Methyl-2-formylnicotinate ≥95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-formylnicotinate

A753076-250MG A753076-5G
77882-643EA 122 USD
77882-643 77882-644
Methyl-2-formylnicotinate ≥95%
Methyl-2-formylnicotinate

Precaución: for Research Use Only

Formula: C₈H₇NO₃
MDL Number: MFCD11052321
CAS Number: 25230-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR