11,12-Didehydro-ε-oxodibenz[b,f]azocine-5(6H)-hexanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: DBCO-C6-acid

A752905-1G A752905-100MG A752905-250MG A752905-50MG
77598-914EA 490.98 USD
77598-914 77622-718 77622-720 77622-722
11,12-Didehydro-ε-oxodibenz[b,f]azocine-5(6H)-hexanoic acid ≥95%
11,12-Didehydro-ε-oxodibenz[b,f]azocine-5(6H)-hexanoic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₉NO₃
MW: 333.3805 g/mol
MDL Number: MFCD22380737
CAS Number: 1425485-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR