PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Phenyl-4,4,5,5-tetramethylimidazoline-1-oxyl 3-oxide, 4,5-Dihydro-4,4,5,5-tetramethyl-2-phenyl-1H-imidazol-1-yloxy-1-oxide, 2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl, PTIO

A752664-250MG A752664-1G
77622-716EA 49.44 USD
77622-716 77622-714
PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) ≥98%
PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl)

Precaución: for Research Use Only

Formula: C₁₃H₁₇N₂O₂
MW: 233.29 g/mol
MDL Number: MFCD00059929
CAS Number: 18390-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR