tert-Butyl 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazine-4(3H)-carboxylate 95+%

Supplier: Ambeed
Warning

A752550-100MG A752550-1G A752550-50MG A752550-250MG
77614-312EA 177.16 USD
77614-312 77614-314 77614-316 77686-759
tert-Butyl 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazine-4(3H)-carboxylate 95+%
tert-Butyl 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazine-4(3H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₉H₂₈BNO₅
MW: 361.24 g/mol
MDL Number: MFCD16036419
CAS Number: 1467057-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR