Ethyl 2-(2-bromothiazol-4-yl)acetate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A752394-250MG A752394-100MG A752394-5G
77684-684EA 84 USD
77684-684 77891-481 77891-482
Ethyl 2-(2-bromothiazol-4-yl)acetate ≥95%, Ambeed.Inc
Ethyl 2-(2-bromothiazol-4-yl)acetate

Precaución: For research use only.

Formula: C₇H₈BrNO₂S
MDL Number: MFCD18413040
CAS Number: 56355-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR