Ferrozine mono-sodium salt hydrate ≥98%

Supplier: Ambeed
Warning

Synonyms: FerroZine iron reagent hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate, 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4''-disulphonic acid monosodium salt hydrate, PDT disulphonate monosodium salt hydrate

A752382-5G A752382-25G
77783-888EA 67 USD
77783-888 77783-887
Ferrozine mono-sodium salt hydrate ≥98%
Ferrozine mono-sodium salt hydrate

Precaución: for Research Use Only

Formula: C₂₀H₁₃N₄NaO₆S₂·H₂O
MW: 492.47 g/mol (anhy)
MDL Number: MFCD00150794
CAS Number: 63451-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR