Mal-PEG6-Acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Mal-peg6-acid

A751859-250MG A751859-100MG
77622-706EA 154.9 USD
77622-706 77783-886
Mal-PEG6-Acid ≥95%
Mal-PEG6-Acid

Precaución: for Research Use Only

Formula: C₁₉H₃₁NO₁₀
MDL Number: MFCD22056307
CAS Number: 518044-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR