23-Hydroxy-3,6,9,12,15,18,21-heptaoxatricosyl 4-methylbenzenesulfonate ≥95%

Supplier: Ambeed
Warning

Synonyms: PEG8-Tos

A751766-250MG A751766-5G
77622-702EA 79.31 USD
77622-702 77622-704
23-Hydroxy-3,6,9,12,15,18,21-heptaoxatricosyl 4-methylbenzenesulfonate ≥95%
23-Hydroxy-3,6,9,12,15,18,21-heptaoxatricosyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₂₃H₄₀O₁₁S
MW: 524.6221 g/mol
MDL Number: MFCD26127806
CAS Number: 886469-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR