PEG6-Tos ≥97%

Supplier: Ambeed
Warning

Synonyms: Peg6-tos, Tos-PEG6-OH

A751716-100MG A751716-250MG
77622-698EA 30.9 USD
77622-698 77622-700
PEG6-Tos ≥97%
PEG6-Tos

Precaución: for Research Use Only

Formula: C₁₇H₂₈O₈S
MW: 392.464 g/mol
MDL Number: MFCD22574780
CAS Number: 42749-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR