m-PEG4-benzaldehyde ≥95%

Supplier: Ambeed
Warning

Synonyms: m-PEG3-0-benzaldehyde, 4-(2-(2-(2-Methoxyethoxy)ethoxy)ethoxy)benzaldehyde

A751689-1G A751689-5G
77573-412EA 172 USD
77573-412 77622-696
m-PEG4-benzaldehyde ≥95%
m-PEG4-benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₅
MDL Number: MFCD22683279
CAS Number: 153364-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR