6-(((tert-Butyldimethylsilyl)oxy)methyl)-2,2,3,3,9,9,10,10-octamethyl-4,8-dioxa-3,9-disilaundecan-6-amine ≥95%

Supplier: Ambeed
Warning

A751366-5G A751366-1G
77595-186EA 307.5 USD
77595-186 77783-883
6-(((tert-Butyldimethylsilyl)oxy)methyl)-2,2,3,3,9,9,10,10-octamethyl-4,8-dioxa-3,9-disilaundecan-6-amine ≥95%
6-(((tert-Butyldimethylsilyl)oxy)methyl)-2,2,3,3,9,9,10,10-octamethyl-4,8-dioxa-3,9-disilaundecan-6-amine

Precaución: for Research Use Only

Formula: C₂₂H₅₃NO₃Si₃
MDL Number: MFCD22574795
CAS Number: 102522-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR