tert-Butyl 14-bromo-3,6,9,12-tetraoxatetradecanoate ≥95%

Supplier: Ambeed
Warning

Synonyms: Br-PEG4-CH2-Boc

A751310-1G A751310-5G A751310-250MG
77602-922EA 198.85 USD
77602-922 77598-898 77783-882
tert-Butyl 14-bromo-3,6,9,12-tetraoxatetradecanoate ≥95%
tert-Butyl 14-bromo-3,6,9,12-tetraoxatetradecanoate

Precaución: for Research Use Only

Formula: C₁₄H₂₇BrO₆
MDL Number: MFCD27977496
CAS Number: 1807505-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR