9,10-Dimethoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium sulfate(2:1) ≥98%

Supplier: Ambeed
Danger

Synonyms: Berberine sulphate Anhydrous

A751297-25G A751297-5G A751297-100MG
77747-482EA 101 USD
77747-482 77747-483 77747-481
9,10-Dimethoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium sulfate(2:1) ≥98%
9,10-Dimethoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium sulfate(2:1)

Precaución: for Research Use Only

Formula: C₄₀H₃₆N₂O₁₂S
MW: 768.79 g/mol
MDL Number: MFCD00031743
CAS Number: 316-41-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR