N-(4-((4-Chlorobenzyl)oxy)pyridin-2-yl)-2-(2,6-difluorophenyl)acetamide 98+%

Supplier: Ambeed
Warning

Synonyms: CPDA

A751287-10MG A751287-5MG
77618-072EA 108.15 USD
77618-072 77618-074
N-(4-((4-Chlorobenzyl)oxy)pyridin-2-yl)-2-(2,6-difluorophenyl)acetamide 98+%
N-(4-((4-Chlorobenzyl)oxy)pyridin-2-yl)-2-(2,6-difluorophenyl)acetamide

Precaución: for Research Use Only

Formula: C₂₀H₁₅ClF₂N₂O₂
MW: 388.79 g/mol
MDL Number: MFCD29472572
CAS Number: 1415834-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR