tert-Butyl 1-bromo-3,6,9,12,15,18-hexaoxahenicosan-21-oate ≥95%

Supplier: Ambeed
Warning

Synonyms: Bromo-PEG6-Boc

A750832-5G A750832-1G A750832-250MG
77573-406EA 313.12 USD
77573-406 77598-894 77622-680
tert-Butyl 1-bromo-3,6,9,12,15,18-hexaoxahenicosan-21-oate ≥95%
tert-Butyl 1-bromo-3,6,9,12,15,18-hexaoxahenicosan-21-oate

Precaución: for Research Use Only

Formula: C₁₉H₃₇BrO₈
MW: 473.3963 g/mol
MDL Number: MFCD22574809
CAS Number: 1393330-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR