26-Hydroxy-3,6,9,12,15,18,21,24-octaoxahexacosyl 4-methylbenzenesulfonate ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tos-PEG9
A750543-1G
A750543-5G
A750543-250MG
A750543-100MG
77783-873EA
78
USD
77783-873
77783-875
77783-874
77783-872
26-Hydroxy-3,6,9,12,15,18,21,24-octaoxahexacosyl 4-methylbenzenesulfonate ≥95%
26-Hydroxy-3,6,9,12,15,18,21,24-octaoxahexacosyl 4-methylbenzenesulfonate
Precaución: for Research Use Only
Formula:
C₂₅H₄₄O₁₂S |
MDL Number:
MFCD27977527 CAS Number: 62573-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...