Mal-PEG2-acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 3-[2-[2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy]ethoxy]propanoic acid, 3-(2-(2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy)ethoxy)propanoic acid

A750400-1G A750400-250MG A750400-100MG
77622-676EA 143.17 USD
77622-676 77622-678 77622-674
Mal-PEG2-acid ≥95%
Mal-PEG2-acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₆
MW: 257.24 g/mol
MDL Number: MFCD23726620
CAS Number: 1374666-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR