tert-Butyl 4,7,10,13,16,19,22-heptaoxapentacos-24-ynoate ≥95%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG7-Boc

A750337-250MG A750337-1G A750337-5G
77598-890EA 127.1 USD
77598-890 77622-672 77783-867
tert-Butyl 4,7,10,13,16,19,22-heptaoxapentacos-24-ynoate ≥95%
tert-Butyl 4,7,10,13,16,19,22-heptaoxapentacos-24-ynoate

Precaución: for Research Use Only

Formula: C₂₂H₄₀O₉
MW: 448.55 g/mol
MDL Number: MFCD22683299
CAS Number: 1818294-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR