Amino-PEG5-acid ≥96%

Supplier: Ambeed
Warning

Synonyms: Amino-PEG5-C2-acid, 1-Amino-3,6,9,12,15-pentaoxaoctadecan-18-oic acid

A750214-50MG A750214-100MG A750214-250MG
77598-888EA 50.23 USD
77598-888 77622-668 77622-670
Amino-PEG5-acid ≥96%
Amino-PEG5-acid

Precaución: for Research Use Only

Formula: C₁₃H₂₇NO₇
MW: 309.356 g/mol
MDL Number: MFCD25424126
CAS Number: 1191078-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR