CBZ-N-amido-PEG4-acid ≥95%

Supplier: Ambeed
Warning

Synonyms: CBZ-N-amido-dPEG®4-acid, 3-Oxo-1-phenyl-2,7,10,13,16-pentaoxa-4-azanonadecan-19-oic acid, Cbz-NH-PEG4-C2-acid

A750065-250MG A750065-5G
77622-666EA 90.64 USD
77622-666 77783-862
CBZ-N-amido-PEG4-acid ≥95%
CBZ-N-amido-PEG4-acid

Precaución: for Research Use Only

Formula: C₁₉H₂₉NO₈
MDL Number: MFCD11041139
CAS Number: 756526-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR