1,3-Di(benzo[d]oxazol-2-yl)benzene ≥98%

Supplier: Ambeed
Warning

A749269-100MG A749269-250MG A749269-1G
77614-304EA 90.64 USD
77614-304 77674-056 77884-673
1,3-Di(benzo[d]oxazol-2-yl)benzene ≥98%
1,3-Di(benzo[d]oxazol-2-yl)benzene

Precaución: for Research Use Only

Formula: C₂₀H₁₂N₂O₂
MDL Number: MFCD00382603
CAS Number: 59049-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR