(S)-2-Amino-5-methylhexanoic acid hydrochloride 98+%

Supplier: Ambeed
Warning

A749231-1G A749231-250MG
77893-477EA 30 USD
77893-477 77893-478
(S)-2-Amino-5-methylhexanoic acid hydrochloride 98+%
(S)-2-Amino-5-methylhexanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO₂
MDL Number: MFCD06795775
CAS Number: 1330286-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR