1,6-Naphthyridine-8-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A748748-1G A748748-100MG A748748-250MG
77614-302EA 642.72 USD
77614-302 77674-050 77674-052
1,6-Naphthyridine-8-carboxylic acid ≥98%
1,6-Naphthyridine-8-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₂
MDL Number: MFCD09863867
CAS Number: 362606-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR