Potassium ((1-(tert-butoxycarbonyl)azetidin-3-yl)methyl)trifluoroborate ≥95%

Supplier: Ambeed
Danger

A748196-1G A748196-250MG
77783-849EA 317 USD
77783-849 77783-850
Potassium ((1-(tert-butoxycarbonyl)azetidin-3-yl)methyl)trifluoroborate ≥95%
Potassium ((1-(tert-butoxycarbonyl)azetidin-3-yl)methyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₉H₁₆BF₃KNO₂
MDL Number: MFCD18712403
CAS Number: 2254447-10-2
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR