(S)-4-(1-(5-Chloro-2-(4-fluorophenoxy)benzamido)ethyl)benzoic acid ≥99%

Supplier: Ambeed
Warning

Synonyms: CJ-42794

A747666-10MG A747666-5MG
77684-677EA 45 USD
77684-677 77684-678
(S)-4-(1-(5-Chloro-2-(4-fluorophenoxy)benzamido)ethyl)benzoic acid ≥99%
(S)-4-(1-(5-Chloro-2-(4-fluorophenoxy)benzamido)ethyl)benzoic acid

Precaución: For research use only.

Formula: C₂₂H₁₇ClFNO₄
MW: 413.83 g/mol
MDL Number: MFCD13184815
CAS Number: 847728-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR