1-(Difluoromethyl)naphthalene 95+%

Supplier: Ambeed
Danger

A747575-1G A747575-250MG
77882-639EA 162 USD
77882-639 77882-640
1-(Difluoromethyl)naphthalene 95+%
1-(Difluoromethyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₁H₈F₂
MDL Number: MFCD12546939
CAS Number: 53731-26-3
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR