1-Cycloheptenylboronic acid pinacol ester ≥98%
Supplier: Ambeed
|
Warning
|
A747196-100MG
A747196-1G
A747196-250MG
77886-695EA
58
USD
77886-695
77886-696
77886-697
1-Cycloheptenylboronic acid pinacol ester ≥98%
1-Cycloheptenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₂₃BO₂ |
MDL Number:
MFCD10700152 CAS Number: 287944-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...