3-Fluoro-1-iodopropane ≥98% stabilised

Supplier: Ambeed
Danger

Synonyms: 1-Fluoro-3-iodopropane

Stabilization: Stabilized with Copper chip

A746952-5G A746952-10G A746952-1G A746952-25G
77614-298EA 20.6 USD
77614-298 77674-046 77674-048 77884-670
3-Fluoro-1-iodopropane ≥98% stabilised
3-Fluoro-1-iodopropane

Precaución: for Research Use Only

Formula: C₃H₆FI
MDL Number: MFCD07784197
CAS Number: 462-40-8
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR