3-Deazaneplanocin A hydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: (1S,2R,5R)-5-(4-Amino-1H-imidazo[4,5-c]pyridin-1-yl)-3-(hydroxymethyl)cyclopent-3-ene-1,2-diol hydrochloride, 3-Deazaneplanocin A HCl

A746301-10MG A746301-1MG A746301-5MG
77684-671EA 452 USD
77684-671 77684-672 77684-673
3-Deazaneplanocin A hydrochloride ≥98%
3-Deazaneplanocin A hydrochloride

Precaución: For research use only.

Formula: C₁₂H₁₅ClN₄O₃
MW: 298.73 g/mol
MDL Number: MFCD18782644
CAS Number: 120964-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR