4-((5-Bromopyridin-2-yl)methyl)morpholine ≥97%
Supplier: Ambeed
|
Warning
|
A746034-1G
A746034-250MG
77891-471EA
127
USD
77891-471
77891-472
4-((5-Bromopyridin-2-yl)methyl)morpholine ≥97%
4-((5-Bromopyridin-2-yl)methyl)morpholine
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BrN₂O |
MDL Number:
MFCD11977594 CAS Number: 294851-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...