Methyl 6-methylpyridazine-3-carboxylate ≥95%

Supplier: Ambeed
Warning

A745720-100MG A745720-1G A745720-250MG
77882-633EA 20 USD
77882-633 77882-634 77882-635
Methyl 6-methylpyridazine-3-carboxylate ≥95%
Methyl 6-methylpyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD13174778
CAS Number: 106584-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR