3-Azaspiro[5.5]undecane-3,9-dicarboxylic acid-3-(tert-butyl) ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-[(tert-butoxy)carbonyl]-3-azaspiro[5.5]undecane-9-carboxylic acid, 3-(tert-Butoxycarbonyl)-3-azaspiro[5.5]undecane-9-carboxylic acid

A745071-100MG A745071-250MG
77614-286EA 90.64 USD
77614-286 77614-288
3-Azaspiro[5.5]undecane-3,9-dicarboxylic acid-3-(tert-butyl) ester ≥98%
3-Azaspiro[5.5]undecane-3,9-dicarboxylic acid-3-(tert-butyl) ester

Precaución: for Research Use Only

Formula: C₁₆H₂₇NO₄
MW: 297.4 g/mol
MDL Number: MFCD11042915
CAS Number: 170228-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR