4-(Trifluoromethoxy)piperidine hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
A744396-100MG
A744396-250MG
77891-469EA
90
USD
77891-469
77891-470
4-(Trifluoromethoxy)piperidine hydrochloride ≥98%
4-(Trifluoromethoxy)piperidine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₁ClF₃NO |
MDL Number:
MFCD23382079 CAS Number: 1612172-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...