(Iodomethyl)cyclobutane ≥98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilized with Copper chip

A744291-1G A744291-5G
77618-050EA 73.13 USD
77618-050 77747-467
(Iodomethyl)cyclobutane ≥98% stabilized
(Iodomethyl)cyclobutane

Precaución: for Research Use Only

Formula: C₅H₉I
MW: 196.029 g/mol
MDL Number: MFCD06410737
CAS Number: 16408-62-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR