(R)-2-Oxoimidazolidine-4-carboxylic acid ≥90%

Supplier: Ambeed
Warning

A743477-1G A743477-250MG
77618-046EA 532.1 USD
77618-046 77747-466
(R)-2-Oxoimidazolidine-4-carboxylic acid ≥90%
(R)-2-Oxoimidazolidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₆N₂O₃
MDL Number: MFCD00272205
CAS Number: 214767-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR