Karanjin ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxy-2-phenyl-4H-furo[2,3-h]chromen-4-one

A743358-10MG A743358-5MG
77670-006EA 158 USD
77670-006 77670-008
Karanjin ≥98%
Karanjin

Precaución: for Research Use Only

Formula: C₁₈H₁₂O₄
MW: 292.28 g/mol
MDL Number: MFCD00017406
CAS Number: 521-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR