3-(3,5-Di-tert-butyl-1-hydroxy-4-oxocyclohexa-2,5-dien-1-yl)propanoic acid ≥95%

Supplier: Ambeed
Warning

A743024-100MG A743024-250MG A743024-50MG
77749-499EA 340 USD
77749-499 77749-500 77749-501
3-(3,5-Di-tert-butyl-1-hydroxy-4-oxocyclohexa-2,5-dien-1-yl)propanoic acid ≥95%
3-(3,5-Di-tert-butyl-1-hydroxy-4-oxocyclohexa-2,5-dien-1-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₆O₄
CAS Number: 83237-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR