Methyl 4-fluoro-1H-pyrrole-2-carboxylate ≥98%

Supplier: Ambeed
Warning

A742887-1G A742887-250MG A742887-100MG A742887-5G
77595-172EA 54.32 USD
77595-172 77674-038 77884-661 77884-662
Methyl 4-fluoro-1H-pyrrole-2-carboxylate ≥98%
Methyl 4-fluoro-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆FNO₂
MDL Number: MFCD11042594
CAS Number: 475561-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR