4-Chloro-5-nitro-1H-pyrrolo[2,3-b]pyridine ≥98%

Supplier: Ambeed
Warning

A742746-100MG A742746-250MG
77873-175EA 62 USD
77873-175 77873-176
4-Chloro-5-nitro-1H-pyrrolo[2,3-b]pyridine ≥98%
4-Chloro-5-nitro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MDL Number: MFCD18072769
CAS Number: 1245645-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR