Methyl-4-chloro-2-fluorobenzoate ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A742601-5G A742601-100G
77684-668EA 22 USD
77684-668 77891-466
Methyl-4-chloro-2-fluorobenzoate ≥98%, Ambeed.Inc
Methyl-4-chloro-2-fluorobenzoate

Precaución: For research use only.

Formula: C₈H₆ClFO₂
MDL Number: MFCD04038392
CAS Number: 148893-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR