2-(3,4-Difluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: Ambeed
Warning

A742343-100MG A742343-250MG A742343-1G
77783-841EA 94 USD
77783-841 77783-843 77783-842
2-(3,4-Difluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(3,4-Difluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₂H₁₄BF₂NO₄
MDL Number: MFCD31653845
CAS Number: 2657617-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR