3,6-Dichlorophthalic anhydride ≥98%

Supplier: Ambeed
Warning

Synonyms: 4,7-Dichloroisobenzofuran-1,3-dione

A741596-1G A741596-5G
77573-376EA 77.25 USD
77573-376 77747-464
3,6-Dichlorophthalic anhydride ≥98%
3,6-Dichlorophthalic anhydride

Precaución: for Research Use Only

Formula: C₈H₂Cl₂O₃
MW: 217.01 g/mol
MDL Number: MFCD00042786
CAS Number: 4466-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR