2,2-Dichloro-1-(3-methyl-2,3-dihydro-4H-benzo[b][1,4]oxazin-4-yl)ethan-1-one ≥98%

Supplier: Ambeed
Danger

Synonyms: Benoxacor

A741554-1G A741554-25G A741554-5G
77669-994EA 23 USD
77669-994 77669-996 77669-998
2,2-Dichloro-1-(3-methyl-2,3-dihydro-4H-benzo[b][1,4]oxazin-4-yl)ethan-1-one ≥98%
2,2-Dichloro-1-(3-methyl-2,3-dihydro-4H-benzo[b][1,4]oxazin-4-yl)ethan-1-one

Precaución: for Research Use Only

Formula: C₁₁H₁₁Cl₂NO₂
MDL Number: MFCD01311820
CAS Number: 98730-04-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR